ContactUsLinkComponent
20X SSPE, liquid, Corning®
20X SSPE, liquid, Corning®
Catalog # MDTC46-021-CM
Supplier:  Corning
CAS Number:  
undefined
20X SSPE, liquid, Corning®
Catalog # MDTC46-021-CM
Supplier:  Corning
CAS Number:  
Specifications
  • Pk:
    6
  • Description:
    20X SSPE, liquid
  • Shipping temperature:
    Ambient
  • Packed:
    6/pack
Specifications
About this item
20X Saline-Sodium-Phosphate-EDTA (SSPE) buffer has applications in nucleic acid hybridization and other molecular biology work.