Order Entry
Switzerland
Orders LinkContactUsLinkComponent
TE buffer, 1X, liquid, Corning®

TE buffer, 1X, liquid, Corning®

1X TE buffer, liquid, 6×1 L, storage temperature: 15 - 30 °C, 7,9 - 8,1, 6
Catalog # MDTC46-009-CM
Supplier:   Corning
TE buffer, 1X, liquid, Corning®
Catalog # MDTC46-009-CM
Supplier:  Corning
CAS Number:  
TE buffer, 1X, liquid, Corning®
TE buffer, 1X, liquid, Corning®

1X TE buffer, liquid, 6×1 L, storage temperature: 15 - 30 °C, 7,9 - 8,1, 6

CAS Number:  
Price
SFr. 361.00
Log in for availability
1 * 6 ST
Product Details & Documents

Tris-EDTA (TE) buffer is commonly used in work with nucleic acids.

Examples include as a solubilisation buffer in DNA and RNA purification, as a running buffer in electrophoresis, and as a general storage buffer for nucleic acids.

Documents

Specifications

Specifications

  • Pk:
    6
  • Description:
    1X TE buffer, liquid, 6×1 L, storage temperature: 15 - 30 °C
  • pH value:
    7,9 - 8,1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.