Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Yeast extract, powder, Corning®

Yeast extract, powder, Corning®

Yeast extract, powder 10000 MW cut-off, 500 g, 500 g
Catalog # MDTC61-231-RO
Supplier:   Corning
Yeast extract, powder, Corning®
Catalog # MDTC61-231-RO
Supplier:  Corning
CAS Number:  
Yeast extract, powder, Corning®

Yeast extract, powder 10000 MW cut-off, 500 g, 500 g

CAS Number:  
Price
310.00 SFr.
Log in for availability
1 * 500 g
Product Details & Documents

Yeast extract is a mixture of amino acids, peptides, water soluble vitamins and carbohydrates obtained from autolysed yeast

This is the water soluble portion with intact B-complex vitamins and can be used as an additive for culture media.

Specifications

Specifications

  • Pk:
    500 g
  • Description:
    Yeast extract, powder 10000 MW cut-off, 500 g
  • Cat. No.:
    MDTC61-231-RO
  • Storage temperature:
    Ambient
  • Shipping temperature:
    Ambient

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.