Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Electrophoresis Buffers
Running Buffers
MOPS buffer, powder, Corning®
MOPS buffer, powder, Corning®
MOPS buffer, powder, Corning®
Catalog # MDTC46-103-RM
Supplier:
Corning
CAS Number:
Certificates
Expand image
MOPS buffer, powder, Corning®
Catalog # MDTC46-103-RM
Supplier:
Corning
CAS Number:
Certificates
Specifications
Pk:
100 g
Description:
MOPS buffer, powder, 100 g
Storage temperature:
Ambient
Shipping temperature:
Ambient
Specifications
About this item
Product Details
Documentation
Product Details
Documentation