ContactUsLinkComponent
Masterflex® L/S® Easy-Load® II Pump Heads with 2-Channels for Precision Tubing, Avantor®
undefined
Masterflex® L/S® Easy-Load® II Pump Heads with 2-Channels for Precision Tubing, Avantor®
  • Gear ratio:
  • 1:1
  • 1:1
  • Max. flow rate per channel:
  • 1000 ml/min
  • 1000 ml/min
  • Min. flow rate per channel:
  • 0,0012 ml/min
  • 0,0012 ml/min
  • No. of channels:
  • 2
  • 2
  • Occlusion:
  • Fixed
  • Fixed
  • Open-head sensor:
  • No
  • No
  • Optimum RPM range:
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • Pump series:
  • L/S
  • L/S
  • Suction lift:
  • 8,8 m (29 ft.)
  • 8,8 m (29 ft.)
  • Tubing retention:
  • Automatic
  • Automatic
  • Tubing sizes accepted:
  • L/S® 13, L/S 14, L/S 16, L/S 25
  • L/S® 13, L/S 14, L/S 16, L/S 25
  • Volume per revolution with L/S 13 tubing:
  • 0,06 ml
  • 0,06 ml
  • Volume per revolution with L/S 14 tubing:
  • 0,21 ml
  • 0,21 ml
  • Volume per revolution with L/S 16 tubing:
  • 0,8 ml
  • 0,8 ml
  • Volume per revolution with L/S 25 tubing:
  • 1,7 ml
  • 1,7 ml
  • Warranty:
  • 2 years
  • 2 years
Frequently Bought Together