ContactUsLinkComponent
Masterflex® Chemically Inert Metering Valves, Avantor®
Masterflex® Chemically Inert Metering Valves, Avantor®
Certificates
undefined
Masterflex® Chemically Inert Metering Valves, Avantor®
Specifications
  • Model:
  • 06394-16
  • 06394-00
  • 06394-02
  • 06394-04
  • Max. pressure:
  • 5,2 bar (75 psi)
  • 5,2 bar (75 psi)
  • 5,2 bar (75 psi)
  • 5,2 bar (75 psi)
  • Max. temperature:
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • Orifice:
  • 6 mm (0,25")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • Wetted materials:
  • PTFE
  • PCTFE and PTFE
  • PCTFE and PTFE
  • PCTFE and PTFE
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year