ContactUsLinkComponent
Masterflex® Flow Ratchet Tubing Control Clamps, Avantor®
Masterflex® Flow Ratchet Tubing Control Clamps, Avantor®
Certificates
undefined
Masterflex® Flow Ratchet Tubing Control Clamps, Avantor®
Specifications
  • Housing material:
  • Polyoxymethylene (POM) Acetal
  • Polyoxymethylene (POM) Acetal
  • Pinch valve tube size:
  • 1/16" to 1/4" OD
  • 1/8" to 1/2" OD
  • Warranty:
  • 3 Months