ContactUsLinkComponent
Micropump Compact Low-Flow Gear Pump Heads - C Mount Design
Micropump Compact Low-Flow Gear Pump Heads - C Mount Design
Certificates
undefined
Micropump Compact Low-Flow Gear Pump Heads - C Mount Design
Specifications
  • Body material:
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • 316 stainless steel
  • Differential pressure:
  • 3,4 bar (50 psi)
  • 5,2 bar (75 psi)
  • 2,8 bar (40 psi)
  • 2,8 bar (40 psi)
  • 1,4 bar (20 psi)
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Gear material:
  • PTFE
  • PPS
  • Carbon graphite
  • Carbon graphite
  • Carbon graphite
  • Inlet connection:
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • Max. temperature:
  • 77 °C (170 °F)
  • 177 °C (350 °F)
  • 177 °C (350 °F)
  • 177 °C (350 °F)
  • 177 °C (350 °F)
  • Max. viscosity:
  • 100 cP
  • 100 cP
  • 100 cP
  • 100 cP
  • 100 cP
  • Min. temperature:
  • –46 °C (–50 °F)
  • –46 °C (–50 °F)
  • –46 °C (–50 °F)
  • –46 °C (–50 °F)
  • –46 °C (–50 °F)
  • Outlet connection:
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • 0,125 NPT(F)
  • Particulates:
  • No
  • No
  • No
  • No
  • No
  • Relief valve:
  • No
  • No
  • No
  • No
  • No
  • Run dry:
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Under 30 seconds
  • Seal material:
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • PTFE
  • Suction lift, wet:
  • 12" H₂O
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year