ContactUsLinkComponent
Masterflex® Union Fittings, Hose Barb, Cross Connector, Avantor®
undefined
Masterflex® Union Fittings, Hose Barb, Cross Connector, Avantor®
  • Model:
  • 31208-55
  • 31407-11
  • 31208-52
  • 31208-53
  • 31208-51
  • 31408-11
  • 31208-54
  • 40726-74
  • 40726-70
  • 40726-73
  • 40726-71
  • Orifice dimensions:
  • 7,143 mm (0,28125")
  • 0,125"
  • 1,588 mm (0,0625")
  • 2,381 mm (0,09375")
  • 1,588 mm (0,0625")
  • 0,125"
  • 3,175 mm (0,125")
  • Sterilisation:
  • Autoclave, gamma irradiation, or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave, gamma irradiation, or ethylene oxide (ETO)
  • Autoclave, gamma irradiation, or ethylene oxide (ETO)
  • Autoclave, gamma irradiation, or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Autoclave, gamma irradiation, or ethylene oxide (ETO)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • Out of box
  • Out of box
  • Out of box
  • Out of box
Frequently Bought Together