ContactUsLinkComponent
Masterflex® L/S® Drive Dual Voltage Power Supply, Avantor®
Masterflex® L/S® Power Supply, Dual Voltage 115/230 VAC to 12 VDC, 12-ft Cable
Catalog # MFLX77200-07
undefined
Masterflex® L/S® Power Supply, Dual Voltage 115/230 VAC to 12 VDC, 12-ft Cable
Catalog # MFLX77200-07
Specifications
  • Description:
    Power supply, dual voltage 115/230 VAC to 12 VDC, 12-ft cable
  • Warranty:
    1 year
  • Compatible with:
    07571-00 L/S drive
  • Pk:
    1
Specifications
About this item
Make the most of your Masterflex drives replacement parts and other accessories.
  • Suitable for use with 07571-00 L/S drive
This power supply is rated for 3.0 Amps at 12 VDC. The total output power is 36 watts at 12 VDC.
  • Description:
    Power supply, dual voltage 115/230 VAC to 12 VDC, 12-ft cable
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Compatible with:
    07571-00 L/S drive
  • Pk:
    1