Corning® cell expansion bags
Anbieter: Corning
Zertifikate
Über diesen Artikel
Corning® Cell Expansion Bags are intended for the expansion and culture of non-adherent cells.
- Made from a single-web polyolefin, gas permeable film
- Port 1 - Two leads with female Luers and pinch clamp
- Port 2 - Dockable fill line with sampling valve and pinch clamp
The integrated tubing allows for functionally closed system filling, feeding, and sampling.
Zugehörige Informationen
Frequently Bought Together