VWR®, Inserts for Screw Neck Vials ND8
Supplier: VWR Collection
Certificates
About this item
Frequently Bought Together
Calcium hypochlorite for synthesis, Sigma-Aldrich®
Supplier: Merck
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR® Septa ND8
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Sterican® Single-Use Needles for Special Applications
Supplier: B. Braun
Screw caps, 9 mm, level 3 high performance applications, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Dichloromethane, EMPARTA® ACS analytical reagent, Supelco®
Supplier: Merck
Lactic acid 90% ≥90% in aqueous solution, GPR RECTAPUR®, pure
Supplier: VWR Chemicals
Interfold and roll hand towels, Tork
Supplier: Essity
Orthophosphoric acid 85% ≥85%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98%
Supplier: VWR Chemicals