Orders LinkContactUsLinkComponent
Compressor-cooled incubators, ICPeco Series
Compressor-cooled incubators, ICPeco Series
Catalog #
Compressor-cooled incubators, ICPeco Series
Catalog #
Supplier:  
Compressor-cooled incubators, ICPeco Series
Product Details & Documents
The ICPeco compressor-cooled incubators are perfect for different ramps for incubating, breeding or storing, ideal for applications that need a larger chamber volume and at the same time quick temperature changes.
  • Powerful with rapid temperature changes
  • Climate-friendly refrigerant (R744)
  • Wider range of applications thanks to cooling down to the freezing point (-12 °C)
  • Temperature ramps programmable via software AtmoCONTROL
  • Intelligent defrost function
These powerful compressor-cooled incubators are available in four different sizes (110, 260, 450 and 750 L). Environmental friendliness is a top priority for the Memmert ICPeco cooled incubators, which is why the climate-friendly refrigerant R744 is used. This is chlorine-free, it is neither toxic nor flammable and has no greenhouse degradation potential. In addition, it improves the refrigerating capacity and has a faster temperature change speed when cooling down compared to units using the conventional refrigerant R134a.
The units have double doors as standard with fully insulated stainless steel and internal glass which prevents contamination and increases or decreases in temperature. At the same time the user has an optimal view on the sensitive load through the wide-area interior glass doors.
The ICPeco models are specifically designed for ramp operation with rapid temperature changes during incubation, breeding or storage in the fields of biology and zoology as well as in the pharmaceutical and food industries.
All Compressor-cooled incubators are supplied with a TwinDISPLAY controller. The TwinDISPLAY offers maximum convenience for complex processes. If the application requires outstanding precision and reliability combined with intuitive operation plus double over-temperature protection including visual alarm the TwinDISPLAY models will provide the functionality, convenience and safety required. Thanks to the intuitive control and AtmoCONTROL logging software even complex processes can be programmed quickly and easily. Multiple overtemperature protection, visual and acoustic alarms as well as the ability to send an automatic alarm message to one or several e-mail addresses will leave the user time for things that really matter.
Certifications: Standard units are safety-approved and bear the EAC test mark.
Ordering information: Supplied without grids or shelves, please order required shelves, stainless steel grids or trays separately. For all chamber volumes a minimum of 2 shelves or grids must be ordered in addition. Range of accessories and factory fitted options such as access ports, interior sockets and lighting are available on request. Please contact Avantor for further details prior to ordering as these can not be retro-fitted.
Caution: For details of the range of shelves and grids, please visit website or contact your local Avantor sales office.
*Note: (not suitable for long-term storing at sub-zero temperatures. During permanent operation, the inner glass door may ice over)