Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Cobalt(II) chloride hexahydrate 97.0-102.0%, GPR RECTAPUR®
 
undefined
 
 
 
Cobalt(II) chloride hexahydrate 97.0-102.0%, GPR RECTAPUR®
 

 

  • Assay (GC):
    97.0 - 102.0%
  • Identification:
    passes test
  • SO4 (Sulphate):
    max. 100 ppm
  • Cu (Copper):
    max. 50 ppm
  • Fe (Iron):
    max. 50 ppm
  • Ni (Nickel):
    max. 0.15%
  • Pb (Lead):
    max. 30 ppm
  • Zn (Zinc):
    max. 0.05%

 

Frequently Bought Together