Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Conductivity meter, SevenDirect SD30 | Avantor

Conductivity meter, SevenDirect SD30

Anbieter:   Mettler - Toledo
Conductivity meter, SevenDirect SD30
Anbieter:  Mettler - Toledo
Zertifikate
Conductivity meter, SevenDirect SD30

3 Varianten

Product Details & Documents

Robust bench top meter for accurate conductivity determinations in the lab and near production.

  • Large colour touch screen provides direct measurement and easy data entry
  • Instructions guide users through the sensor calibration process
  • Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)

The large colour touch screen offers a direct menu access, fast data entry and guides users through routine tasks. The instruments supports GLP compliance by storing time stamp, sample, sensor and method IDs with every measurement.

Spezifikationen für diese Produktfamilie

Technische Daten

  • Catalog No:
  • 662-2391
  • 662-2408
  • 662-2410
  • Model:
  • Conductivity meter SevenDirect SD30
  • Conductivity meter SevenDirect SD30 kit
  • Conductivity meter SevenDirect SD30 Pure H₂O kit
  • Conductivity range:
  • 0,000 μS/cm - 2000 mS/cm
  • 0,000 μS/cm - 2000 mS/cm
  • 0,000 μS/cm - 2000 mS/cm
  • Conductivity resolution:
  • 0,001 - 1
  • 0,001 - 1
  • 0,001 - 1
  • Conductivity accuracy:
  • 0,5% of measured value
  • 0,5% of measured value
  • 0,5% of measured value
  • Resistivity range:
  • 0,00 - 100,0 MΩ*cm
  • 0,00 - 100,0 MΩ*cm
  • 0,00 - 100,0 MΩ*cm
  • Salinity range:
  • 0,00 - 80,00 psu
  • 0,00 - 80,00 psu
  • 0,00 - 80,00 psu
  • TDS range:
  • 0.00 mg/L – 1000 g/L
  • 0.00 mg/L – 1000 g/L
  • 0.00 mg/L – 1000 g/L
  • Temperature range:
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 (0...100 °C)
  • ±0,1 (0...100 °C)
  • ±0,1 (0...100 °C)
  • Calibration points:
  • 1 or 2
  • 1 or 2
  • 1 or 2
  • Calibration:
  • Linear, Manual cell constant
  • Linear, Manual cell constant
  • Linear, Manual cell constant

Technische Daten

Product Family Options

Produktinformationen

  • Description
    Verfügbarkeit
    Preis
  • Conductivity meter SevenDirect SD30
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST - SFr. 2’065.00
    SFr. 2’065.00
    1 * 1 ST
       SFr. 2’065.00

    Spezifikationen:
    Mehr
    Specifications
    Cat. No.
    662-2391
    Model
    Conductivity meter SevenDirect SD30
    Conductivity range
    0,000 μS/cm - 2000 mS/cm
    Conductivity resolution
    0,001 - 1
    Conductivity accuracy
    0,5% of measured value
    Resistivity range
    0,00 - 100,0 MΩ*cm
    Salinity range
    0,00 - 80,00 psu
    TDS range
    0.00 mg/L – 1000 g/L
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 (0...100 °C)
    Calibration points
    1 or 2
    Calibration
    Linear, Manual cell constant
  • Conductivity meter SevenDirect SD30 kit
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST - SFr. 2’665.00
    SFr. 2’665.00
    1 * 1 ST
       SFr. 2’665.00

    Spezifikationen:
    Mehr
    Specifications
    Cat. No.
    662-2408
    Model
    Conductivity meter SevenDirect SD30 kit
    Conductivity range
    0,000 μS/cm - 2000 mS/cm
    Conductivity resolution
    0,001 - 1
    Conductivity accuracy
    0,5% of measured value
    Resistivity range
    0,00 - 100,0 MΩ*cm
    Salinity range
    0,00 - 80,00 psu
    TDS range
    0.00 mg/L – 1000 g/L
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 (0...100 °C)
    Calibration points
    1 or 2
    Calibration
    Linear, Manual cell constant
  • Conductivity meter SevenDirect SD30 Pure H₂O kit
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 1 ST
    1 * 1 ST - SFr. 2’950.00
    SFr. 2’950.00
    1 * 1 ST
       SFr. 2’950.00

    Spezifikationen:
    Mehr
    Specifications
    Cat. No.
    662-2410
    Model
    Conductivity meter SevenDirect SD30 Pure H₂O kit
    Conductivity range
    0,000 μS/cm - 2000 mS/cm
    Conductivity resolution
    0,001 - 1
    Conductivity accuracy
    0,5% of measured value
    Resistivity range
    0,00 - 100,0 MΩ*cm
    Salinity range
    0,00 - 80,00 psu
    TDS range
    0.00 mg/L – 1000 g/L
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 (0...100 °C)
    Calibration points
    1 or 2
    Calibration
    Linear, Manual cell constant
Accessories

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2413
    Electrode arm, EasyPlace Complete supports efficient measurement and calibration; With perfect vertical movement, the risk of any spillage is minimised; Includes parking beaker, parking beaker insert and sachet holder
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 352.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2382
    Replaceable protective cover for SevenDirect meters
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 58.30

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2383
    Screws and feet set EasyPlace
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 31.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2414
    Magnetic stirrer, EasyMix for use with SevenDirect meters; The EasyMix can be controlled manually or automatically by the SevenDirect meter
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 417.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2415
    Parking beaker, EasyPlace electrode arm; Fill it with storage solution and keep your electrode perfectly conditioned also in short breaks between measurements; or use it to store the wetting cap while measuring
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 26.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2416
    Parking beaker insert, EasyPlace; should be used while using the parking beaker with storage solution
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 9.80

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2417
    Sachet holder, EasyPlace rotating; hold up to three buffer sachets and prevent spillage; It can be rotated to place one buffer after the other in the calibration position
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 50.30

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2418
    Sensor holder, EasyPlace Single for the EasyPlace electrode arm holds one sensor
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 26.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2419
    Sensor Holder, EasyPlace Triple for the EasyPlace electrode arm holds up to three sensors
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 31.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2420
    Sensor Holder, EasyPlace Single Extended for the EasyPlace electrode arm with extended length for use with large sample vessels. Holds one sensor
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 31.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2421
    Pole EasyPlace, 30 cm, standard pole for the EasyPlace electrode arm
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 251.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2422
    Pole, EasyPlace 50 cm, extended pole for the EasyPlace electrode arm for use with large sample vessels
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 302.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2423
    Base plate for the EasyPlace electrode arm
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 151.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2424
    Power supply, 18 W, does not include country-specific plug (order plugs separately with model (662-2426)
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 100.50

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2425
    Top cover pole, EasyPlace or the top of the EasyPlace pole, has two positions for long-term sensor storage
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 26.00

Produktinformationen

  • Katalognr.
    Description
    Verfügbarkeit
    Preis
  • 662-2426
    Plug set, set of country-specific plugs for Power supply 18 W (662-2424)
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 34.10

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.