Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

pH meter, SevenDirect SD20

pH meter, SevenDirect SD20
 
pH meter, SevenDirect SD20
pH meter, SevenDirect SD20

 

 

Robust bench top meter for accurate pH determinations in the lab and near production.

  • Large colour touch screen provides direct measurement and easy data entry
  • Instructions guide users through the sensor calibration process
  • Stores data, such as time stamp, sample and sensor ID with every measurement and prints or exports records automatically
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP 54 in-use)

The large colour touch screen offers a direct menu access, fast data entry and guides users through routine tasks. The instruments supports GLP compliance by storing time stamp, sample, sensor and method IDs with every measurement.

 

 

  • Catalog No:
  • 662-2390
  • 662-2394
  • 662-2396
  • 662-2402
  • 662-2397
  • 662-2398
  • 662-2399
  • 662-2395
  • 662-2400
  • pH range:
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • −2,000...+20,000
  • pH resolution:
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • 0,1 / 0,01 / 0,001
  • pH accuracy:
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • ±0,002
  • mV range:
  • −2000,0...+2000,0
  • −2000,0...+2000,0
  • −2000,0...+2000,0
  • −2000,0...+2000,0
  • −2000,0...+2000,0
  • −2000,0...+2000,0
  • −2000,0...+2000,0
  • −2000,0...+2000,0
  • −2000,0...+2000,0
  • mV resolution:
  • 1 / 0,1
  • 1 / 0,1
  • 1 / 0,1
  • 1 / 0,1
  • 1 / 0,1
  • 1 / 0,1
  • 1 / 0,1
  • 1 / 0,1
  • 1 / 0,1
  • mV accuracy:
  • ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
  • ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
  • ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
  • ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
  • ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
  • ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
  • ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
  • ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
  • ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
  • Temperature range:
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • −30,0…+130,0 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ±0,1 °C (0...100 °C)
  • ±0,1 °C (0...100 °C)
  • ±0,1 °C (0...100 °C)
  • ±0,1 °C (0...100 °C)
  • ±0,1 °C (0...100 °C)
  • ±0,1 °C (0...100 °C)
  • ±0,1 °C (0...100 °C)
  • ±0,1 °C (0...100 °C)
  • ±0,1 °C (0...100 °C)
  • Calibration:
  • Linear, Segmented
  • Linear, Segmented
  • Linear, Segmented
  • Linear, Segmented
  • Linear, Segmented
  • Linear, Segmented
  • Linear, Segmented
  • Linear, Segmented
  • Linear, Segmented
  • Calibration points:
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • 1 - 5
  • 1 - 5

 

Product Family Options

 

  • Description
     
     
  • pH meter SevenDirect SD20
       
       
    1 * 1 ST
    1 * 1 ST - 1 475.00 SFr.
    1 475.00 SFr.
    1 * 1 ST
       1 475.00 SFr.

     
     
    Specifications
    Cat. No.
    662-2390
    pH range
    −2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    mV range
    −2000,0...+2000,0
    mV resolution
    1 / 0,1
    mV accuracy
    ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C (0...100 °C)
    Calibration
    Linear, Segmented
    Calibration points
    1 - 5
  • pH meter SevenDirect SD20 kit
       
       
    1 * 1 ST
    1 * 1 ST - 1 950.00 SFr.
    1 950.00 SFr.
    1 * 1 ST
       1 950.00 SFr.

     
     
    Specifications
    Cat. No.
    662-2394
    pH range
    −2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    mV range
    −2000,0...+2000,0
    mV resolution
    1 / 0,1
    mV accuracy
    ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C (0...100 °C)
    Calibration
    Linear, Segmented
    Calibration points
    1 - 5
  • pH meter SevenDirect SD20 HA kit
       
       
    1 * 1 ST
    1 * 1 ST - 2 065.00 SFr.
    2 065.00 SFr.
    1 * 1 ST
       2 065.00 SFr.

     
     
    Specifications
    Cat. No.
    662-2396
    pH range
    −2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    mV range
    −2000,0...+2000,0
    mV resolution
    1 / 0,1
    mV accuracy
    ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C (0...100 °C)
    Calibration
    Linear, Segmented
    Calibration points
    1 - 5
  • pH meter SevenDirect SD20 LowVol kit
       
       
    1 * 1 ST
    1 * 1 ST - 2 305.00 SFr.
    2 305.00 SFr.
    1 * 1 ST
       2 305.00 SFr.

     
     
    Specifications
    Cat. No.
    662-2402
    pH range
    −2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    mV range
    −2000,0...+2000,0
    mV resolution
    1 / 0,1
    mV accuracy
    ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C (0...100 °C)
    Calibration
    Linear, Segmented
    Calibration points
    1 - 5
  • pH meter SevenDirect SD20 Organic kit
       
       
    1 * 1 ST
    1 * 1 ST - 2 250.00 SFr.
    2 250.00 SFr.
    1 * 1 ST
       2 250.00 SFr.

     
     
    Specifications
    Cat. No.
    662-2397
    pH range
    −2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    mV range
    −2000,0...+2000,0
    mV resolution
    1 / 0,1
    mV accuracy
    ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C (0...100 °C)
    Calibration
    Linear, Segmented
    Calibration points
    1 - 5
  • pH meter SevenDirect SD20 Pure H₂O kit
       
       
    1 * 1 ST
    1 * 1 ST - 2 265.00 SFr.
    2 265.00 SFr.
    1 * 1 ST
       2 265.00 SFr.

     
     
    Specifications
    Cat. No.
    662-2398
    pH range
    −2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    mV range
    −2000,0...+2000,0
    mV resolution
    1 / 0,1
    mV accuracy
    ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C (0...100 °C)
    Calibration
    Linear, Segmented
    Calibration points
    1 - 5
  • pH meter SevenDirect SD20 Solids kit
       
       
    1 * 1 ST
    1 * 1 ST - 2 185.00 SFr.
    2 185.00 SFr.
    1 * 1 ST
       2 185.00 SFr.

     
     
    Specifications
    Cat. No.
    662-2399
    pH range
    −2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    mV range
    −2000,0...+2000,0
    mV resolution
    1 / 0,1
    mV accuracy
    ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C (0...100 °C)
    Calibration
    Linear, Segmented
    Calibration points
    1 - 5
  • pH meter SevenDirect SD20 Stirrer kit
       
       
    1 * 1 ST
    1 * 1 ST - 2 290.00 SFr.
    2 290.00 SFr.
    1 * 1 ST
       2 290.00 SFr.

     
     
    Specifications
    Cat. No.
    662-2395
    pH range
    −2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    mV range
    −2000,0...+2000,0
    mV resolution
    1 / 0,1
    mV accuracy
    ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C (0...100 °C)
    Calibration
    Linear, Segmented
    Calibration points
    1 - 5
  • pH meter SevenDirect SD20 Viscous kit
       
       
    1 * 1 ST
    1 * 1 ST - 2 375.00 SFr.
    2 375.00 SFr.
    1 * 1 ST
       2 375.00 SFr.

     
     
    Specifications
    Cat. No.
    662-2400
    pH range
    −2,000...+20,000
    pH resolution
    0,1 / 0,01 / 0,001
    pH accuracy
    ±0,002
    mV range
    −2000,0...+2000,0
    mV resolution
    1 / 0,1
    mV accuracy
    ±0,1 (−500,0...+500,0 mV) / ±0,2 (−2000,0...+2000,0 mV)
    Temperature range
    −30,0…+130,0 °C
    Temperature resolution
    0,1 °C
    Temperature accuracy
    ±0,1 °C (0...100 °C)
    Calibration
    Linear, Segmented
    Calibration points
    1 - 5
Accessories

 

  •  
    Description
     
     
  • 662-2382
    Replaceable protective cover for SevenDirect meters
       
    1 * 1 ST - SFr. 58.30

 

  •  
    Description
     
     
  • 662-2383
    Screws and feet set EasyPlace
       
    1 * 1 ST - SFr. 31.00

 

  •  
    Description
     
     
  • 662-2414
    Magnetic stirrer, EasyMix for use with SevenDirect meters; The EasyMix can be controlled manually or automatically by the SevenDirect meter
       
    1 * 1 ST - SFr. 417.00

 

  •  
    Description
     
     
  • 662-2415
    Parking beaker, EasyPlace electrode arm; Fill it with storage solution and keep your electrode perfectly conditioned also in short breaks between measurements; or use it to store the wetting cap while measuring
       
    1 * 1 ST - SFr. 26.00

 

  •  
    Description
     
     
  • 662-2418
    Sensor holder, EasyPlace Single for the EasyPlace electrode arm holds one sensor
       
    1 * 1 ST - SFr. 26.00

 

  •  
    Description
     
     
  • 662-2419
    Sensor Holder, EasyPlace Triple for the EasyPlace electrode arm holds up to three sensors
       
    1 * 1 ST - SFr. 31.00

 

  •  
    Description
     
     
  • 662-2420
    Sensor Holder, EasyPlace Single Extended for the EasyPlace electrode arm with extended length for use with large sample vessels. Holds one sensor
       
    1 * 1 ST - SFr. 31.00

 

  •  
    Description
     
     
  • 662-2421
    Pole EasyPlace, 30 cm, standard pole for the EasyPlace electrode arm
       
    1 * 1 ST - SFr. 251.00

 

  •  
    Description
     
     
  • 662-2425
    Top cover pole, EasyPlace or the top of the EasyPlace pole, has two positions for long-term sensor storage
       
    1 * 1 ST - SFr. 26.00

 

  •  
    Description
     
     
  • 662-2426
    Plug set, set of country-specific plugs for Power supply 18 W (662-2424)
       
    1 * 1 ST - SFr. 34.10

 

  •  
    Description
     
     
  • 662-2413
    Electrode arm, EasyPlace Complete supports efficient measurement and calibration; With perfect vertical movement, the risk of any spillage is minimised; Includes parking beaker, parking beaker insert and sachet holder
       
    1 * 1 ST - SFr. 352.00

 

  •  
    Description
     
     
  • 662-2416
    Parking beaker insert, EasyPlace; should be used while using the parking beaker with storage solution
       
    1 * 1 ST - SFr. 9.80

 

  •  
    Description
     
     
  • 662-2417
    Sachet holder, EasyPlace rotating; hold up to three buffer sachets and prevent spillage; It can be rotated to place one buffer after the other in the calibration position
       
    1 * 1 ST - SFr. 50.30

 

  •  
    Description
     
     
  • 662-2422
    Pole, EasyPlace 50 cm, extended pole for the EasyPlace electrode arm for use with large sample vessels
       
    1 * 1 ST - SFr. 302.00

 

  •  
    Description
     
     
  • 662-2423
    Base plate for the EasyPlace electrode arm
       
    1 * 1 ST - SFr. 151.00

 

  •  
    Description
     
     
  • 662-2424
    Power supply, 18 W, does not include country-specific plug (order plugs separately with model (662-2426)
       
    1 * 1 ST - SFr. 100.50