Order Entry
Switzerland
ContactUsLinkComponent
Hydroxylammonium chloride ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent (max. 0.01 ppm Hg)
Health
Irritant
Corrosive
Environment
Hydroxylammonium chloride ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent (max. 0.01 ppm Hg)
Catalog # 24709.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Corrosive
Environment
Hydroxylammonium chloride ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent (max. 0.01 ppm Hg)
Catalog # 24709.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay :
    min. 99.0%
  • Solubility in alcohol:
    passes test
  • Free acidity:
    max 25 meq/g
  • pH (20°C; 5%):
    2,5 - 3,5
  • Heavy metals (as Pb):
    max. 5 ppm
  • Ignition residue (SO4):
    max. 100 ppm
  • Insolubility in water:
    max. 50 ppm
  • NH4 (Ammonium):
    max. 0.2%
  • SO4 (Sulphate):
    max. 20 ppm
  • As (Arsenic):
    max. 1 ppm
  • Cu (Copper):
    max. 10 ppm
  • Fe (Iron):
    max. 20 ppm
  • Hg (Mercury):
    max. 10 ppb
  • Pb (Lead):
    max . 5 ppm
  • Conforms to Reag.Ph.Eur.:
    passes test

Specification Test Results

About this item

 :
 :69.49 g/mol
Merck Index:13,04853
Boiling Pt:306 °C (1013 hPa)
Melting Pt:151 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-798-2
UN:2923
ADR:8 (6.1),III
 :GHS08
 :GHS07
 :GHS05
 :GHS09
CAS Number:5470-11-1
Formula:NH₂OH·HCl
MDL Number:MFCD00051089