Stand-up lectern tables, 304 stainless steel
Supplier: TEKNOMEK
Certificates
About this item
304 grade stainless steel lecterns with sloped top that are perfect for standing at. Durable and strong, with hygienic design features to minimise risk. They are fully-welded, with reduce dirt traps to reduce risk. Choose from freestanding, wall mounted or mobile, with a wide choice of shelf options too.
- 304-grade EU sourced stainless steel
- Supplied fully welded, no construction or assembly required
- Ideal for standing
- Inclined top with lip to hold documents in place
- Hygienic design throughout
- Brushed dull polish finish
- Choice of mobile or static
- Choice of no shelf, 1 shelf or 2 shelves
- Minimised dirt traps
Sturdy 30 mm box section frame, designed with hygiene in mind, all dirt traps minimised from every angle.
Load capacity: 150 kg
Frequently Bought Together