Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Arched tip forceps

Arched tip forceps

Stainless steel, 100 mm, Arched, Straight, 1,3×3×0,2 mm
Catalog # FSCI11071-10
Marketsource
Supplier:   FINE SCIENCE TOOLS
Arched tip forceps
Catalog # FSCI11071-10
Supplier:  FINE SCIENCE TOOLS
Marketsource
Arched tip forceps
Arched tip forceps

Stainless steel, 100 mm, Arched, Straight, 1,3×3×0,2 mm

Price
SFr. 103.06
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Length:
    100 mm
  • Material:
    Stainless steel
  • Tip style:
    Arched
  • Tip size:
    1,3×3×0,2 mm
  • Tip shape:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.