Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Circle forceps

Circle forceps

Stainless steel, 115 mm, Straight, Ø 0,7 mm
Catalog # FSCI11072-11
Marketsource
Supplier:   FINE SCIENCE TOOLS
Circle forceps
Catalog # FSCI11072-11
Supplier:  FINE SCIENCE TOOLS
Marketsource
Circle forceps
Circle forceps

Stainless steel, 115 mm, Straight, Ø 0,7 mm

Price
SFr. 22.90
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Length:
    115 mm
  • Material:
    Stainless steel
  • Tip size:
    Ø 0,7 mm
  • Tip shape:
    Straight
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.