Dumont #5, mirror finish forceps
Supplier: FINE SCIENCE TOOLS
Certificates
About this item
Frequently Bought Together
Tubing clamps, stainless steel
Supplier: USBECK Laborgeräte
VWR®, Test Sieves, 200×50 mm
Supplier: VWR Collection
SS-3/8-DISC-2, Standard Aperture - 2 Mic 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
SCISSORS LAB WITH BUTTON 125MM 1 * 1 items
Supplier: RSG
Dumont #5SF forceps
Supplier: FINE SCIENCE TOOLS
VWR®, Clamps
Supplier: VWR Collection
Scoops
Supplier: USBECK Laborgeräte
1 1/4 SCREWS FOR 48 WELL ALUMINUM REACTION PLATE
Supplier: MARKET SOURCE PART PROCESS
Test sieves, 100×40 mm
Supplier: Retsch GmbH
Scalpels
Supplier: BOCHEM
Pots, laboratory, without lid
Supplier: BOCHEM
General purpose spoons
Supplier: RSG
Magnetic stirring bars, PTFE, cylindrical, with centre ring
Supplier: Brand
Stainless Steel Bottles, GL45
Supplier: Sampling Systems
Test sieves
Supplier: Retsch GmbH
Implantable pico-Drive: 2x4x7 mm 3.5 mm travel fiber optic compatible 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
Rotilabo-chain clamps 1 * 1 items
Supplier: Roth Carl