Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dumont #5-45 forceps

Dumont #5-45 forceps

#5/45, Dumoxel, 110 mm, Standard, Angled (45°), 0,1×0,06 mm
Catalog # FSCI11251-35
Marketsource
Supplier:   FINE SCIENCE TOOLS
Dumont #5-45 forceps
Catalog # FSCI11251-35
Supplier:  FINE SCIENCE TOOLS
Marketsource
Dumont #5-45 forceps
Dumont #5-45 forceps

#5/45, Dumoxel, 110 mm, Standard, Angled (45°), 0,1×0,06 mm

Price
SFr. 37.56
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Type:
    #5/45
  • Length:
    110 mm
  • Material:
    Dumoxel
  • Tip style:
    Standard
  • Tip size:
    0,1×0,06 mm
  • Tip shape:
    Angled (45°)