ContactUsLinkComponent
Dumont #7, fine forceps
Dumont #7, fine forceps
Catalog # FSCI11274-20
Supplier:  FINE SCIENCE TOOLS
undefined
Dumont #7, fine forceps
Catalog # FSCI11274-20
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Type:
    #7
  • Length:
    115 mm
  • Material:
    Inox
  • Tip style:
    Biology
  • Pk:
    1
  • Tip size:
    0,07×0,03 mm
  • Tip shape:
    Curved