ContactUsLinkComponent
Dumont #L5, sliding lock forceps
Dumont #L5, sliding lock forceps
Catalog # FSCI11253-27
Supplier:  FINE SCIENCE TOOLS
undefined
Dumont #L5, sliding lock forceps
Catalog # FSCI11253-27
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Type:
    #L5
  • Length:
    110 mm
  • Material:
    Inox
  • Tip style:
    Standard
  • Pk:
    1
  • Tip size:
    0,1×0,06 mm
  • Tip shape:
    Straight
Specifications
About this item
Plastic clamping ring holds tips together and can be locked in place. Fully autoclavable.