Orders LinkContactUsLinkComponent
Dumont SS forceps
Marketsource
Dumont SS forceps
Marketsource
Dumont SS forceps
Dumont SS forceps
Product Family Options
  • Type
    Material
    Length
    Tip style
    Tip shape
    Tip size
  • SS
    Inox
    135 mm
    Standard
    Angled (45°)
    0,2×0,12 mm
    1 * 1 ST
    1 * 1 ST - SFr. 38.48
    SFr. 38.48
    1 * 1 ST
       SFr. 38.48
    Marketsource
    Marketsource
  • SS
    Inox
    135 mm
    Standard
    Straight
    0,2×0,12 mm
    1 * 1 ST
    1 * 1 ST - SFr. 34.81
    SFr. 34.81
    1 * 1 ST
       SFr. 34.81
    Marketsource
    Marketsource