ContactUsLinkComponent
Fine forceps, broad shanks
Fine forceps, broad shanks
Catalog # FSCI11445-12
Supplier:  FINE SCIENCE TOOLS
undefined
Fine forceps, broad shanks
Catalog # FSCI11445-12
Supplier:  FINE SCIENCE TOOLS
Specifications
  • Type:
    Fine
  • Length:
    120 mm
  • Material:
    Stainless steel
  • Tip style:
    Broad shanks
  • Pk:
    1
  • Tip size:
    0,2×0,12 mm
  • Tip shape:
    Straight