Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Students splinter forceps

Students splinter forceps

Stainless steel, 105 mm, Serrated, Straight, 0,3 mm
Catalog # FSCI91156-11
Marketsource
Supplier:   FINE SCIENCE TOOLS
Students splinter forceps
Catalog # FSCI91156-11
Supplier:  FINE SCIENCE TOOLS
Marketsource
Students splinter forceps
Students splinter forceps

Stainless steel, 105 mm, Serrated, Straight, 0,3 mm

Price
SFr. 17.69
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Length:
    105 mm
  • Material:
    Stainless steel
  • Tip style:
    Serrated
  • Tip width:
    0,3 mm
  • Tip shape:
    Straight