Order Entry
Switzerland
ContactUsLinkComponent
Enterococcus faecium WDCM 00010 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Enterococcus faecium WDCM 00010 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000105-10EA
Supplier:  Merck
CAS Number:  
Enterococcus faecium WDCM 00010 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Catalog # VT000105-10EA
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10
  • Description:
    Enterococcus faecium WDCM 00010 Vitroids™, 1000 - 10000 CFU mean value range
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    –20 °C
  • Product type:
    Hydrosoluble discs

Specifications

About this item

Derived from CECT® strains,

  • Ready-to-use microbiological controls minimize the need for maintaining control strains in the test laboratory
  • Defined CFU range and low standard deviation
  • Fast, reliable and easy to use
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Caution: CECT is a registered trademark of Universitat de Valencia.