Order Entry
Switzerland
ContactUsLinkComponent
Micro needle holder with suture cutter
Micro needle holder with suture cutter
  FSCI12075-14
undefined
Micro needle holder with suture cutter
  FSCI12075-14

 

  • Description:
    Clamping length of 5 mm
  • Length:
    140 mm
  • Material:
    Stainless steel
  • Tip style:
    Scissors
  • Pk:
    1
  • Tip width:
    0,8 mm
  • Tip shape:
    Curved