Orders LinkContactUsLinkComponent
Student standard pattern forceps
Student standard pattern forceps
Marketsource
Supplier:   FINE SCIENCE TOOLS
Student standard pattern forceps
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Student standard pattern forceps
Student standard pattern forceps
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Material
    Length
    Tip style
    Tip shape
    Tip width
    Availability
    Price
  • Stainless steel
    115 mm
    Serrated
    Straight
    2,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12.37 SFr.
    12.37 SFr.
    1 * 1 ST
       12.37 SFr.
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    145 mm
    Serrated
    Straight
    3,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13.04 SFr.
    13.04 SFr.
    1 * 1 ST
       13.04 SFr.
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    160 mm
    Serrated
    Straight
    3,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12.83 SFr.
    12.83 SFr.
    1 * 1 ST
       12.83 SFr.
    Specifications:
    Marketsource
    Marketsource