Orders LinkContactUsLinkComponent
Tissue forceps, 1×2 teeth
Tissue forceps, 1×2 teeth
Marketsource
Supplier:   FINE SCIENCE TOOLS
Tissue forceps, 1×2 teeth
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Tissue forceps, 1×2 teeth
Tissue forceps, 1×2 teeth
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Material
    Length
    Tip style
    Tip shape
    Tip size
    Availability
    Price
  • Stainless steel
    140 mm
    1×2 teeth
    Curved
    3×2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 21.53
    SFr. 21.53
    1 * 1 ST
       SFr. 21.53
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    120 mm
    1×2 teeth
    Straight
    1,7×1,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 15.57
    SFr. 15.57
    1 * 1 ST
       SFr. 15.57
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    145 mm
    1×2 teeth
    Straight
    2×1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 14.66
    SFr. 14.66
    1 * 1 ST
       SFr. 14.66
    Specifications:
    Marketsource
    Marketsource
  • Stainless steel
    155 mm
    1×2 teeth
    Straight
    2,5×1,7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 16.03
    SFr. 16.03
    1 * 1 ST
       SFr. 16.03
    Specifications:
    Marketsource
    Marketsource