Titanium micro serrefines
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
About this item
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- MaterialLengthTip styleTip shapeTip sizeAvailabilityPrice
- FSCI18555-04Strong clamp press, 125 gStrong clamp press, 125 gTitanium13 mmSerratedStraight4×0,75 mmLog in for availability1 * 1 STSFr. 121.84Specifications:MarketsourceMarketsource
- FSCI18555-03Medium clamp press, 100 gMedium clamp press, 100 gTitanium15 mmSerratedStraight6×1 mmLog in for availability1 * 1 STSFr. 121.84Specifications:MarketsourceMarketsource
- FSCI18555-02Light clamp press, 85 gLight clamp press, 85 gTitanium16 mmSerratedStraight8×2 mmLog in for availability1 * 1 STSFr. 121.84Specifications:MarketsourceMarketsource
- FSCI18555-01Light clamp press, 60 gLight clamp press, 60 gTitanium19 mmSerratedStraight10×2 mmLog in for availability1 * 1 STSFr. 121.84Specifications:MarketsourceMarketsource