ContactUsLinkComponent
Bacillus subtilis subsp. spizizenii WDCM 00003 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
  VT000036-10EA
 :  Merck
 :  
Bacillus subtilis subsp. spizizenii WDCM 00003 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
  VT000036-10EA
 :  Merck
 :  
  • Pk:
    10
  • Description:
    Bacillus subtilis subsp. spizizenii WDCM 00003 Vitroids™, 1000 - 10000 CFU mean value range
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    −20 °C
  • Product type:
    Hydrosoluble discs
Derived from CECT® strains.
 : This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.