ContactUsLinkComponent
Bacillus cereus NCTC 7464 Lenticule® Discs Suitable for Microbiology, Certified Reference Material, Supelco®
  CRM07464L-10EA
 :  Merck
 :  
Bacillus cereus NCTC 7464 Lenticule® Discs Suitable for Microbiology, Certified Reference Material, Supelco®
  CRM07464L-10EA
 :  Merck
 :  
  • Pk:
    10
  • Description:
    Bacillus cereus NCTC 7464 Lenticule® discs, 30-120 CFU
  • M-Clarity quality level:
    MQ300
  • Storage temperature:
    −20 °C
  • Product type:
    Hydrosoluble discs
 : This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
 : LENTICULE® discs are manufactured by Sigma-Aldrich under licence from Public Health England.