Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Kaye scissors, extra large loops

Blunt/blunt, curved tips, large loops, 105 mm, 22 mm, +
Catalog # FSCI14171-10
Marketsource
Supplier:   FINE SCIENCE TOOLS
Kaye scissors, extra large loops
Catalog # FSCI14171-10
Supplier:  FINE SCIENCE TOOLS
Marketsource
Kaye scissors, extra large loops
Kaye scissors, extra large loops

Blunt/blunt, curved tips, large loops, 105 mm, 22 mm, +

Price
SFr. 81.99
Log in for availability
1 * 1 ST
Product Details & Documents

These scissors are made of stainless steel.

Specifications
  • Type:
    Blunt/blunt, curved tips, large loops
  • Length:
    105 mm
  • Serrated:
    +
  • Blade length:
    22 mm