Orders LinkContactUsLinkComponent
Fine scissors, Tungsten Carbide
Fine scissors, Tungsten Carbide
Marketsource
Supplier:   FINE SCIENCE TOOLS
Fine scissors, Tungsten Carbide
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Fine scissors, Tungsten Carbide
Fine scissors, Tungsten Carbide
4 Options Available Below
Product Details & Documents
These scissors are made of stainless steel with tungsten carbide inserts.
Product Family Options
Product Information
  • Type
    Length
    Blade length
    Availability
    Price
  • Sharp/sharp, straight tips, tungsten carbide
    90 mm
    22 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 74.95
    SFr. 74.95
    1 * 1 ST
       SFr. 74.95
    Specifications:
    Marketsource
    Marketsource
  • Sharp/sharp, curved tips, tungsten carbide
    90 mm
    22 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 81.94
    SFr. 81.94
    1 * 1 ST
       SFr. 81.94
    Specifications:
    Marketsource
    Marketsource
  • Sharp/sharp, straight tips, tungsten carbide
    115 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 84.73
    SFr. 84.73
    1 * 1 ST
       SFr. 84.73
    Specifications:
    Marketsource
    Marketsource
  • Sharp/sharp, curved tips, tungsten carbide
    115 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 91.25
    SFr. 91.25
    1 * 1 ST
       SFr. 91.25
    Specifications:
    Marketsource
    Marketsource