Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Moria Fine scissors

Sharp/sharp, straight tips, 105 mm, 25 mm
  FSCI14370-22
Marketsource
Moria Fine scissors
  FSCI14370-22
Marketsource
Moria Fine scissors
Moria Fine scissors

Sharp/sharp, straight tips, 105 mm, 25 mm

 
97.11 SFr.
   
1 * 1 ST
 

These scissors are made of inox.

 
  • Type:
    Sharp/sharp, straight tips
  • Length:
    105 mm
  • Seq:
    01
  • Blade length:
    25 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.