Über diesen Artikel
Disposable funnel for reliable filling of liquids, ideal for filling bottles and other containers with a narrow filler opening.
- For easy and reliable filling or filtering
- Suitable for use in laboratories and clean rooms or for sampling
- Cleanroom manufactured (cleanroom Class 7)
- Individually packaged and optionally sterilised
- Complies with EU food requirements and FDA regulations
The disposable funnel ensures no loss when transferring liquid samples or free-flowing solids. That makes this practical and hygienic pouring aid a helpful accessory for sampling or in laboratories.
Cleanroom produced funnel with suspension eye and venting fins for ventilation to ensure uniform flow while pouring. In combination with a filter paper the disposable funnel is also well suited for filtering.
The disposable funnel for liquids is ideal for use in laboratories and clean rooms or for sampling. It can be used in pharmaceuticals, chemistry and cosmetics and the food and animal feed industries, as well as all areas where special purity is important.
Technische Daten
- Catalog No:
- 221-0314
- 221-0304
- 221-0303
- 221-0305
- 221-0306
- 221-0307
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact, Sustainable Materials
- Low Manufacturing Impact, Sustainable Materials
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 Certificate
- ISO 14001 Certificate
- ISO 14001 Certificate
- ACT-Label
- ACT-Label
- ISO 14001 Certificate