Order Entry
Switzerland
ContactUsLinkComponent
Acetonitrile for LC-MS
Flammable
Irritant
Acetonitrile for LC-MS
Catalog # HEWLG2453-85050
CAS Number:  
undefined
Flammable
Irritant
Acetonitrile for LC-MS
Catalog # HEWLG2453-85050
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :41.05 g/mol
Density:0.78
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
 :GHS02
 :GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878