Imager model organism package, THUNDER
Catalog # 630-3394
Supplier: LEICA MICROSYSTEMS
Specifications
- Description:Leica® THUNDER imager model organism package
- Pk:1 SET
Specifications
About this item
The THUNDER imager model organism allows fast and easy 3-D exploration of whole organisms (e.g., Drosophila, C. elegans, zebrafish, plants and mice) for developmental or molecular biology research. Eliminate the hassle of out-of-focus blur while maintaining the capabilities and ease of use typical for Leica stereo microscopes.
Takes research to the next level with improved image quality to get the most information from your precious model organisms with THUNDER imager model organism.
Certifications: CE and CAS.