ContactUsLinkComponent
Olympus® safety helmet, EVO®5
Olympus® safety helmet, EVO®5
Catalog # JSPAAKF170-000-100
Supplier:  JSP
undefined
Olympus® safety helmet, EVO®5
Catalog # JSPAAKF170-000-100
Supplier:  JSP
Specifications
  • Description:
    EVO®5 Olympus® Safety helmet
  • Pk:
    1
Specifications
About this item
The EVO®5 Olympus® safety helmet combines a super strong ABS shell designed for superior all day protection with the comfort benefits of the Evolution® 3D Adjustment™ harness system. The helmet passes all relevant industrial international standards.
  • Global multi-standard hard hat
  • Tough ABS shell
  • Vented design
  • 6-point Terylene cradle harness system
  • Revolution® wheel ratchet adjustment
The EVO®5 Olympus® safety helmet features a 6-point terylene cradle harness system and an Egyptian cotton Chamlon™ sweatband that offers unrivalled comfort. 3-D adjustment offers 1-2-3 point harness depth settings for a perfect fit. The Revolution® wheel ratchet has deep nape strap that enables the firmest retention of any helmet, with a simple turn fastening system.
Side and rear ventilation for optimum air movement inside the shell, reducing helmet temperatures by an average 2 to 3 degrees, increasing the comfort for the wearer. The textile harness features unique 1-2-3 point harness depth settings. This design provides the most precise fit attainable on an industrial safety helmet.
Certifications: EN 397; ANSI / ISea Z89.1; CSA Z94.1; GB2811; AS/NZS1801.
Eyewear for safety helmets
Product Information