Sweatband, Chamlon™
Catalog # JSPAAJA840-000-200
Supplier: JSP
Specifications
- Description:Replacement Chamlon™ Sweatband
- Pk:10
Specifications
About this item
Replacement Chamlon™ sweatband.
- For use with the EVO® range of safety helmets
- pH neutral
- Dermatologically tested
Replacement Chamlon sweatband for use with the EVO® range of safety helmets. Egyptian cotton core with porous PU coating for maximum sweat absorption.