ContactUsLinkComponent
Switch & Go 8 Hour Multi Plug PAPR, Jetstream®, JSP LTD
Switch & Go 8 Hour Multi Plug PAPR, Jetstream®, JSP LTD
Catalog # JSPACBB610-211-100
Supplier:  JSP
undefined
Switch & Go 8 Hour Multi Plug PAPR, Jetstream®, JSP LTD
Catalog # JSPACBB610-211-100
Supplier:  JSP
Specifications
  • Description:
    Jetstream® Switch & Go 8 Hour multi plug PAPR
  • Pk:
    1
Specifications
About this item
Jetstream® is a belt-mounted rechargeable respirator with a large capacity disposable dust filter. Jetstream® delivers 180 L/min. of air through the filter via a highly flexible hose to the desired headtop.
  • Belt-mounted rechargeable respirator
  • Large capacity disposable dust filter
  • No face fit required
  • PSL filter
The Jetstream® waist belt and power unit weighs just 765g. The Switch & Go unit comes complete with a white nylon head top. The system offers an assigned protection factor (APF) of 20 x Workplace Exposure Limit (WEL) for very fine dusts, fibres, fumes, and aqueous mists.
Certifications: EN 12941:1998.
Delivery: Jetstream® Switch & Go pack contains: PSL filter, hose, charger, carry case, battery, white nylon headtop, flow meter, and multi-region plug.