Orders LinkContactUsLinkComponent
Safety spectacles, Eiger™
Safety spectacles, Eiger™
Supplier:   JSP
Safety spectacles, Eiger™
Supplier:  JSP
Certificates
Safety spectacles, Eiger™
Safety spectacles, Eiger™
3 Options Available Below
Product Details & Documents
Sporty design with optimal curved lens and large field of vision.
  • Moulded browguard
  • Excellence coating
  • Optimal lateral coverage
Active style and sporty design. Unique Flexofit technology engineered into temple cushioning to enhance wearer comfort, and to ensure a non-slip, low pressure fit.
Excellence coating on the optimal curved lens, large field of vision, moulded browguard. Additional gasket and strap accessory pack available to deliver a perfect dust barrier.
Certifications: Meets EN 166:2001.
Product Family Options
Product Information
  • Colour
    Lenses
    Availability
    Price
  • JSPA1EIGGREEN23CKN
    Eiger™ safety spectacles
    Eiger™ safety spectacles
    Black/green
    Clear
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 9.30
    SFr. 9.30
    1 * 1 ST
       SFr. 9.30
  • JSPA1EIGGREEN23SKN
    Eiger™ safety spectacles
    Eiger™ safety spectacles
    Black/green
    Smoke
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 9.30
    SFr. 9.30
    1 * 1 ST
       SFr. 9.30
  • JSPA1EIGLWP23CKN
    Eiger™ safety spectacles
    Eiger™ safety spectacles
    Purple
    Clear
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 6.50
    SFr. 6.50
    1 * 1 ST
       SFr. 6.50