ContactUsLinkComponent
Grade 0860 ½ qualitative filter papers, Whatman™
Grade 0860 ½ qualitative filter papers, Whatman™
Certificates
undefined
Grade 0860 ½ qualitative filter papers, Whatman™
Specifications
  • Catalog No:
  • 512-0525
  • 512-0526
  • 512-0524
  • WHAT10334607
  • Grade:
  • 0860 ½
  • 0860 ½
  • 0860 ½
  • 0860 ½
  • Weight:
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • 75 g/m²
  • Thickness:
  • 170 µm
  • 170 µm
  • 170 µm
  • 170 µm
  • Particle retention:
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • 7 - 12 µm
  • Filtration speed:
  • 60 s (Herzberg)
  • 60 s (Herzberg)
  • 60 s (Herzberg)
  • 60 s (Herzberg)