Order Entry
Switzerland
ContactUsLinkComponent
VWR®, DNA extraction Kit for Real-time PCR food samples analysis
VWR®, DNA extraction Kit for Real-time PCR food samples analysis
Catalog # M04018BF
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, DNA extraction Kit for Real-time PCR food samples analysis
Catalog # M04018BF
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Refine target molecule:
    DNA preps
  • Sample type:
    Food samples
  • Target molecule:
    DNA
  • Description:
    DNA extraction kit for Real-time PCR food samples analysis
  • Storage conditions:
    0…4 ºC
  • Details:
    2 bottles; for 100 samples

Specifications

About this item

Rapid method for DNA extraction and purification from all types of microorganisms in a variety of matrices for molecular biology.

  • Rapid method for DNA extraction
  • 100 samples for VWR® Real-time PCR food samples kit

VWR Real-Time PCR kits for detection and quantification of Legionella and Salmonella spp.

Product Information