Recombinant Human Glutathione Peroxidase 2
Supplier: SELENOZYME
Certificates
About this item
The human GPX2 selenoprotein is encoded by the GPX2 gene and has potent hydrogen peroxide reducing activity using glutathione.
We provide the human GPX2 enzyme with approx. 20% Sec contents, mixed with inactive forms containing Sec-to-Lys and Sec-to-Gln substitutions, at more than 95% purity in aliquots at amounts from 0,5 to 5 units.
- Genuine Sec-containing selenoproteins
- No animals used as protein sources (grown in bacteria)
- More than 90% Sec contents
- More than 95% purity
Delivery: Protein stored in 25 mM Tris-HCl (pH 8.0) with 250 mM KCl, 2 mM 2-Mercaptoethanol and 10% glycerol. Concentration 1 mg protein/ml. Shipped in cold pack by courier service. Store at −20 °C upon delivery.
Packaging: Aliquots
Caution: Centrifuge the tube briefly before opening the lid.
Specifications
- Storage conditions:Protein stored in 25 mM Tris-HCl (pH 8.0) with 250 mM KCl, 2 mM 2-Mercaptoethanol and 10% glycerol
- Purity:≥ 95%
- Concentration:Concentration 1 mg protein/ml buffer.
Specifications
Frequently Bought Together