Recombinant C. elegans thioredoxin reductase 1
Supplier: SELENOZYME
Certificates
About this item
The TrxR enzyme in C. elegans is the single selenoprotein in this organism, exerting a wide range of functions in redox control and for antioxidant systems. It is also together with glutathione reductase essential for the molting process.
We provide the main isoform of the C. elegans TrxR enzyme with more than 90% Sec contents and more than 95% purity in aliquots at amounts from 5 to 50 units.
- Genuine Sec-containing selenoproteins
- No animals used as protein sources (grown in bacteria)
- More than 90% Sec contents
- More than 95% purity
Delivery: Protein stored in TE buffer with 50% glycerol. Concentration 1 mg protein/ml. Shipped in cold pack by courier service. Store at −20 °C upon delivery.
Packaging: Aliquots
Caution: Centrifuge the tube briefly before opening the lid.
Specifications
- Storage conditions:Protein stored in Tris-EDTA buffer with 50% glycerol.
- Purity:≥ 95%
- Concentration:Concentration 1 mg protein/ml buffer.
Specifications
Frequently Bought Together