VWR®, Cryoboxes, Half Size, Cardboard
Supplier: VWR Collection
Certificates
About this item
Cardboard, coated.
- Waterproof coating
- Freezable to –140 °C
- Dividers available separately
- Available in five colours
For storage of cryovials at low temperatures.
Frequently Bought Together