Order Entry
Switzerland
ContactUsLinkComponent
Safety spectacles, uvex i-lite planet
Safety spectacles, uvex i-lite planet
Catalog # 111-2574
Supplier:  UVEX
   Sustainable Materials
   Low Manufacturing Impact
   Product Transparency and Disclosure
   Sustainable Packaging
   Energy Efficient
undefined
Safety spectacles, uvex i-lite planet
Catalog # 111-2574
Supplier:  UVEX
   Sustainable Materials
   Low Manufacturing Impact
   Product Transparency and Disclosure
   Sustainable Packaging
   Energy Efficient

Specifications

  • Type:
    Anti-UV (UV 400), anti-scratch, anti-fog, uvex supravision excellence
  • Colour:
    Anthracite
  • Environmentally Preferable:
    Sustainable Materials,
    Low Manufacturing Impact,
    Product Transparency and Disclosure,
    Sustainable Packaging,
    Energy Efficient
  • Environmental certification:
    Production plant: ISO 50001
  • Lenses:
    Clear PA
  • Pk:
    1

Specifications

About this item

These metal-free, versatile 3-in-1 protective eyewear range allows a flexible choice for many applications.

  • Large, flat lens design for an unrestricted field of vision
  • UV400 - 100% protection against dangerous UVA and UVB radiation to 400 nm
  • Sustainably produced using bio-based materials

The robust polycarbonate lens with proven uvex supravision coating technology with permanent anti-fog and scratch-resistant features. x-tended sideshield provides additional side protection. The multi-stage adjustable side arms allow a high degree of customisation in order to guarantee a good fit on any head or face shape. Ergonomically shaped side arms for a perfect fit without creating pressure points.

Frame colour: Anthracite, turquoise.

Certifications: Compliant with: EN 166, EN 170. Frame: W EN 166 F CE. Lens: 2C-1.2 W 1 FKN CE.